Learn More
Selleck Chemical LLC ABBV-744
SDP
Supplier:  Selleck Chemical LLC S872310MM/1ML
Encompass_Preferred
ABBV-744 is a BDII-selective BET bromodomain inhibitor that inhibits BRD2 BRD3 and BRD4 It is developed for treating AML and cancers
Catalog No. 50-000-01347