Learn More
Medchemexpress LLC HY-145923 10mg , ABBV-CLS-484 CAS:2489404-97-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14592310MG
Encompass_Preferred
HY-145923 10mg ABBV-CLS-484 CAS: 2489404-97-7 ABBV-CLS-484 is a potent PTPN1 or PTPN2 inhibitor with a sub-nanomolar activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0872