Learn More
Selleck Chemical LLC Abemaciclib mesylate
SDP
Supplier:  Selleck Chemical LLC S71581G
Encompass_Preferred
Abemaciclib mesylate is a potent and selective inhibitor of CDK4 and CDK6 with IC50 of 2 nM and 10 nM in cell-free assays respectively Phase 3
Catalog No. 50-000-04106