Learn More
TARGETMOL CHEMICALS INC ABEMACICLIB METABOLITE M 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T107385MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Abemaciclib metabolite M20 (CDK4/6-IN-4) is an active metabolite of Abemaciclib. Abemaciclib metabolite M20 is a selective CDK4/6 inhibitor and can be used for researches on the treatment of cancer. purity: 98%
Catalog No. 50-258-1678