Learn More
Selleck Chemical LLC Abemaciclib 25mg 1231929-97-7 LY2835219
SDP
Supplier:  Selleck Chemical LLC S571625MG
Encompass_Preferred
Abemaciclib (LY2835219) is a cell cycle inhibitor selective for CDK4/6 with IC50 of 2 nM and 10 nM in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1644728