Learn More
Medchemexpress LLC Abequolixron (hydrochloride) | 610318-54-2 | 99.9% | 596.08 g·mol⁻¹ | C34H33ClF3NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY111498A5MG
Encompass_Preferred
RGX-104 (abequolixron) is an orally bioavailable small-molecule liver X receptor (LXR) agonist used in preclinical and research settings to modulate innate immunity through transcriptional activation of the ApoE gene. Supplied as a characterized research-grade solid, it is suitable for assay development and pharmacological evaluation in vitro and in vivo.
  • Orally bioavailable LXR agonist suitable for pharmacology studies.
  • Modulates innate immunity via ApoE transcriptional activation.
  • Provided as characterized research-grade solid for assay development.
  • Analytical data available, including molecular formula, molecular weight, and purity.
  • Compatible with in vitro and in vivo experimental models.
Catalog No. 50-002-63504