Learn More
Biolegend Purified (azide-free) anti-β-Amyloid, 1-16
Supplier:  Biolegend 803004
Encompass_Preferred
Purified (azide-free) anti-β-Amyloid, 1-16 [6E10]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, Direct ELISA, IHC-P, IHC-F, EM; Size: 25 uL
Catalog No. NC1421533