Learn More
ABclonal Technology ABflo 488 anti-Mouse F4/80 mAb
SDP
Supplier:  ABclonal Technology A27479100T
Encompass_Preferred
Predicted to enable G protein-coupled receptor activity Predicted to be involved in adenylate cyclase-activating G protein-coupled receptor signaling pathway Predicted to act upstream of or within G protein-coupled receptor signaling pathway and adaptive immune response Located in external side of plasma membrane Is expressed in several structures including cardiovascular system central nervous system genitourinary system hemolymphoid system and intestine Orthologous to human ADGRE1 (adhesion G protein-coupled receptor E1)
Catalog No. 50-000-13257