Learn More
ABclonal Technology ABflo 488 Rabbit anti- CD28
SDP
Supplier:  ABclonal Technology A25056200T
Encompass_Preferred
Enables protein kinase binding activity Acts upstream of or within several processes including positive regulation of immune response positive regulation of intracellular signal transduction and regulation of lymphocyte activation Located in external side of plasma membrane and immunological synapse Human ortholog(s) of this gene implicated in multiple sclerosis and type 1 diabetes mellitus Orthologous to human CD28 (CD28 molecule)
Catalog No. 50-000-12836