Learn More
ABclonal Technology ABflo 488 Rat anti-Mouse CD3
SDP
Supplier:  ABclonal Technology A27161500T
Encompass_Preferred
This gene is predicted to enable identical protein binding and transmembrane signaling receptor activity playing a role in positive thymic T cell selection It is part of the alpha-beta T cell receptor complex and expressed in the thymus primordium Human orthologs are implicated in immunodeficiency 19 and severe combined immunodeficiency The gene is involved in nervous system development positive regulation of cell adhesion T cell activation cytokine production and signal transduction It is located in cellular components such as dendritic spines plasma membrane and immunological synapse The gene is expressed in the colon and hemolymphoid system Its human orthologs are implicated in immunodeficiency 18 The gene is orthologous to human CD3D and CD3E
Catalog No. 50-000-12273