Learn More
ABclonal Technology ABflo 594 Rabbit anti- CD47
SDP
Supplier:  ABclonal Technology A24379200T
Encompass_Preferred
Predicted to enable protein binding activity involved in heterotypic cell-cell adhesion and thrombospondin receptor activity Involved in regulation of nitric oxide biosynthetic process Acts upstream of or within several processes including monocyte aggregation opsonization and positive regulation of phagocytosis Located in extracellular exosome Is integral component of plasma membrane Is expressed in several structures including alimentary system central nervous system genitourinary system hemolymphoid system gland and liver and biliary system Orthologous to human CD47 (CD47 molecule)
Catalog No. 50-000-09799