Learn More
ABclonal Technology ABflo 647 Rabbit anti-Dog CD27
SDP
Supplier:  ABclonal Technology A22576500T
Encompass_Preferred
The protein encoded by this gene is a member of the TNF-receptor superfamily This receptor is required for generation and long-term maintenance of T cell immunity It binds to ligand CD70 and plays a key role in regulating B-cell activation and immunoglobulin synthesis This receptor transduces signals that lead to the activation of NF-kappaB and MAPK8 JNK Adaptor proteins TRAF2 and TRAF5 have been shown to mediate the signaling process of this receptor CD27-binding protein (SIVA) a proapoptotic protein can bind to this receptor and is thought to play an important role in the apoptosis induced by this receptor
Catalog No. 50-000-07754