Learn More
ABclonal Technology ABflo 647 Rabbit anti- S100A9
SDP
Supplier:  ABclonal Technology A26916500T
Encompass_Preferred
Predicted to enable calcium ion binding activity and calcium-dependent protein binding activity Involved in peptidyl-cysteine S-nitrosylation positive regulation of blood coagulation and regulation of translation Acts upstream of or within several processes including actin cytoskeleton reorganization astrocyte development and regulation of integrin biosynthetic process Predicted to be located in cell junction cytosol and nucleoplasm Predicted to be active in cytoplasm extracellular space and nucleus Is expressed in several structures including jaw liver otic capsule skeleton and spleen red pulp Human ortholog(s) of this gene implicated in myocarditis Orthologous to human S100A9 (S100 calcium binding protein A9)
Catalog No. 50-000-11131