Learn More
Aviva Systems Biology Corp ABHD6 Antibody - C-terminal region (ARP65850_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65850_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABHD6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human ABHD6. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-580