Learn More
Cayman Chemical AbIrateron Acetate-d4 10mg
SDP
Supplier:  Cayman Chemical 2209610 MG
Encompass_Preferred
An internal standard for the quantification of abiraterone acetate by GC- or LC-MS
Catalog No. 50-298-6998