Learn More
Medchemexpress LLC Abiraterone | 154229-19-3 | 99.8% | 200 MG
SDP
Supplier:  Medchemexpress LLC HY70013200MG
Encompass_Preferred
Abiraterone is a potent and irreversible CYP17A1 inhibitor with antiandrogen activity. It inhibits both the 17α-hydroxylase and 17,20-lyase activity of the cytochrome p450 enzyme CYP17. It is for research use only.
  • Potent and irreversible CYP17A1 inhibitor
  • Exhibits antiandrogen activity
  • Inhibits 17α-hydroxylase with IC50 of 2.5 nM
  • Inhibits 17,20-lyase activity with IC50 of 15 nM
Catalog No. 50-004-33795