Learn More
Medchemexpress LLC HY-70013 5g , Abiraterone CB-7598 CAS:154229-19-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY700135G
Encompass_Preferred
Medchemexpress, HY-70013 5g Abiraterone CB-7598 CAS:154229-19-3 Formula:C24H31NO IC50: 2 to 4 nM (CYP17) Purity:98% Abiraterone is a potent, selective, and irreversible CYP17 inhibitor with IC 50 of 2 to 4 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2223