Learn More
Selleck Chemical LLC Abiraterone S1123-25mg
SDP
Supplier:  Selleck Chemical LLC S112325MG
Encompass_Preferred
Abiraterone is a potent CYP17 inhibitor with IC50 of 2 nM in a cell-free assay Abiraterone (CB-7598) is an androgen biosynthesis inhibitor
Catalog No. 50-310-8529