Learn More
Medchemexpress LLC Abiraterone decanoate | 2486052-18-8 | 99.7% | 503.76 g/mol | C34H49NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1457865MG
Encompass_Preferred
Abiraterone decanoate is an intramuscular prodrug of abiraterone that provides controlled release of the active agent and produces long-acting CYP17 inhibition. It is supplied as a high-purity research compound suitable for preclinical laboratory studies.
  • Provides controlled release of active abiraterone via intramuscular delivery.
  • Delivers long-acting CYP17 inhibition for sustained pharmacologic effect.
  • High purity suitable for preclinical and laboratory research.
  • Available in a 5 mg pack for small-scale studies.
  • Soluble in DMSO for formulation and dosing preparations.
  • Datasheet and safety data sheet available to support use and handling.
Catalog No. 50-002-21032