Learn More
Medchemexpress LLC Abiraterone decanoate | 2486052-18-8 | 99.7% | C34H49NO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14578650MG
Encompass_Preferred
Abiraterone decanoate is a potent Abiraterone proagent that provides controlled release of Abiraterone and long-acting CYP17 inhibition via intramuscular (IM) delivery. This solid, white to off-white laboratory chemical is intended for research use only.
  • Potent abiraterone proagent
  • Provides controlled release of abiraterone
  • Offers long-acting CYP17 inhibition
  • Suitable for intramuscular (IM) delivery
  • Targets CYP17
  • Appears as a white to off-white solid
  • Used in the manufacture of substances
Catalog No. 50-003-55280