Learn More
Medchemexpress LLC Abiraterone sulfate | 1993430-25-3 | 98.9% | 429.57 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14837510MM/1ML
Encompass_Preferred
Abiraterone sulfate is a metabolite of Abiraterone. Abiraterone is a potent and irreversible CYP17A1 inhibitor with antiandrogen activity. It is intended for research use only.
  • Metabolite of Abiraterone
  • Potent and irreversible CYP17A1 inhibitor
  • Antiandrogen activity
  • For research use only
Catalog No. 50-003-61313