Learn More
Agilent Technologies ABLE COMPETENT CELL KIT
Supplier:  Agilent Technologies 200170
Encompass
ABLE Competent Cell Kit. Clone DNA encoding toxic proteins, Reduce plasmid copy number, increasing cell viability, Contain F' episome allowing M13 infection. Contains: 5 x 0.2mL ABLE C and 5 x 0.2mL ABLE K cells
Catalog No. 50-125-026