Learn More
Cayman Chemical Abnormal Cannabidiol
SDP
Supplier:  Cayman Chemical 100042595 MG
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; not found in Cannabis; has been used as an internal standard for the detection of cannabinoids in hemp extracts; intended for research and forensic applications
Catalog No. 50-288-7642