Learn More
TARGETMOL CHEMICALS INC ABR-238901 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T391155MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABR-238901 is an oral active S100A8/A9 blocker that inhibits the interaction of S100A8/A9 with its receptors RAGE(receptor for advanced glycation end products) and TLR4 (toll-like receptor 4). ABR-238901 has potential as a compound for the treatment of myocardial infarction (MI). purity: 99%
Catalog No. 50-258-0067