Learn More
Medchemexpress LLC HY-141537 5mg , ABR-238901 CAS:1638200-22-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1415375MG
Encompass_Preferred
HY-141537 5mg ABR-238901 CAS: 1638200-22-2 ABR-238901 is an orally active and potent S100A8/A9 blocker and inhibits S100A8/A9 interaction with its receptors RAGE (receptor for advanced glycation endproducts) and TLR4 (toll-like receptor 4). ABR-238901 has the potential for myocardial infarction (MI) research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1839