Learn More
Electron Microscopy Sciences Flexible Shaft Machine Abrasive Band Arbor, 1/2"
SDP
Supplier:  Electron Microscopy Sciences 6215112
Encompass_Preferred
Abrasive band arbors have rubber heads on 3/32" (2.35mm) shank.
Diameter: 1/2"
Catalog No. 50-243-84