Learn More
Electron Microscopy Sciences Flexible Shaft Machine Abrasive Band Coarse Grit, 1/4"
SDP
Supplier:  Electron Microscopy Sciences 6215012
Encompass_Preferred
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 1/4"
Catalog No. 50-243-83