Learn More
Medchemexpress LLC Abrocitinib | 1622902-68-4 | MFCD30187577 | 99.3% | 323.41 g·mol⁻¹ | C14H21N5O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10742925MG
Encompass_Preferred
Abrocitinib is a potent, orally active, and selective Janus kinase 1 (JAK1) inhibitor used in preclinical pharmacology. It shows strong JAK1 inhibition (reported IC50 ≈ 29 nM) with reduced activity against JAK2 and TYK2, and is used in in vitro and in vivo models of autoimmune and inflammatory disease.
  • Potent and selective JAK1 inhibition.
  • Demonstrated cellular activity inhibiting STAT phosphorylation.
  • High purity for reproducible experimental results.
  • Available as solid and as DMSO solution for flexible dosing.
  • Multiple in vivo formulation protocols provided for administration.
  • Recommended storage conditions support long-term stability.
Catalog No. 50-002-39873