Learn More
Medicus Health ABS Tube Rack - 50mL, 100 Place
SDP
Supplier:  Medicus Health 7260M1/Q020526CG2
Encompass
• ABS Tube Rack - 50mL, 100 Place allows you to store a large volume of centrifuge tubes in one organized rack
• Lightweight, durable ABS
• Safe for refrigerated storage
• Excellent for storing 50mL centrifuge tubes
• Tubes sold separately
• Overall dimemnsions: 14.4"L x 14.4"W x 3"H
Catalog No. NC3932072