Learn More
Medchemexpress LLC Bmy 7378 | 21102-95-4 | MFCD00153771 | 99.9% | 458.42 | C22H33Cl2N3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10055450MG
Encompass_Preferred
BMY 7378 is a small-molecule research compound that acts as a selective antagonist of the α1D-adrenergic receptor and a partial agonist at the 5-HT1A receptor. It is supplied as a solid for in vitro receptor-binding and pharmacological studies, and is characterized by molecular formula C22H33Cl2N3O3 and molecular weight 458.42 g/mol.
  • Selective antagonist of α1D-adrenergic receptor.
  • Partial agonist at the 5-HT1A receptor.
  • High purity (99.92%).
  • Solid, white to off-white appearance for standard laboratory handling.
  • Supplied in calibrated milligram pack sizes for quantitative use.
  • Recommended storage: 4°C sealed; in solvent keep at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-13454