Learn More
Medchemexpress LLC 2'-3'-cyclic GMP-AMP (2',3'-cGAMP) | 1441190-66-4 | 100.0% | 674.41 g/mol | C20H24N10O13P2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10056410MG
Encompass_Preferred
2',3'-cGAMP is an endogenous cyclic dinucleotide that binds STING and potently induces interferon-β. Supplied as a high-purity solid research reagent, it is intended for in vitro and in vivo studies of the cGAS-STING pathway.
  • High purity: 99.95%.
  • Potent STING agonist that induces interferon-β.
  • Chemical formula C20H24N10O13P2; molecular weight 674.41 g/mol.
  • Supplied as a solid; common pack size 10 mg.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-12788