Learn More
Medchemexpress LLC Abscisic acid-d6 | 721948-65-8 | MFCD30725553 | 98.7% | 270.36 g/mol | C15H14D6O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY100560S1MG
Encompass_Preferred
Abscisic acid-d6 is the deuterium-labeled analogue of abscisic acid supplied as a high-purity reference standard for analytical and biochemical research. It is intended for use as an internal standard or tracer in GC-MS and LC-MS assays and retains the biological activity profile of native abscisic acid. CAS 721948-65-8. For research use only; not for human or clinical use.
  • Deuterium-labeled analogue (d6) for mass spectrometry internal standards.
  • High purity suitable for quantitative analytical methods.
  • Small 1 mg package for trace-level standard preparation.
  • Compatible with GC-MS and LC-MS workflows.
  • Retains biological activity profile of native abscisic acid for comparative studies.
  • Provided for research use only; not for clinical use.
Catalog No. 50-002-12333