Learn More
Medchemexpress LLC ABT-072 (potassium trihydrate) | 1132940-31-8 | 99.6% | 561.69 | C24H32KN3O8S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY101634A10MM 1ML
Encompass_Preferred
ABT-072 (potassium trihydrate) is the potassium trihydrate salt form of a potent, orally active non-nucleoside hepatitis C virus (HCV) NS5B polymerase inhibitor. It is intended for research and analytical use and is reported to exhibit strong antiviral potency (HCV GT1a EC50 = 1 nM; HCV GT1b EC50 = 0.3 nM). The material is supplied with supporting documentation for safe handling and characterization.
  • Potent NS5B polymerase inhibition at low nanomolar concentrations.
  • Supplied as a ready-to-use 10 mM solution in a small vial.
  • High reported purity suitable for analytical applications.
  • Provided with SDS and COA for safety and characterization.
  • Suitable for in vitro antiviral and preclinical studies.
Catalog No. 50-002-47366