Learn More
TARGETMOL CHEMICALS INC ABT 102 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T295225MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABT 102 (CHEMBL398338) is a selective antagonist of transient receptor potential vanilloid 1 (TRPV1) receptor. purity: 99%
Catalog No. 50-258-0454