Learn More
Apexbio Technology LLC ABT 1614-12-6 50mg
SDP
Supplier:  Apexbio Technology LLC B773750
Encompass_Preferred
ABT (1-Aminobenzotriazole CAS 1614-12-6) is a non-selective irreversible inhibitor of cytochrome P450 enzymes (CYP450) It inactivates P450 enzymes by covalently binding to the heme moiety resulting in permanent loss of enzymatic activity ABT is widely employed in pharmacokinetic and drug metabolism studies to investigate metabolic pathways mediated by CYP450 delineate drug-drug interactions and distinguish between P450-dependent and independent metabolic mechanisms This compound serves as a valuable tool for characterizing xenobiotic metabolism during preclinical drug development
Catalog No. 50-000-87785