Learn More
Apexbio Technology LLC ABT-199 1257044-40-8 10mg
SDP
Supplier:  Apexbio Technology LLC A819410
Encompass_Preferred
ABT-199 (CAS 1257044-40-8) is a selective small-molecule inhibitor targeting BCL-2 developed through structure-guided drug design By specifically interfering with the mitochondrial apoptotic pathway regulated by BCL-2 ABT-199 induces apoptosis predominately in cells dependent on BCL-2 signaling rather than BCL-XL In preclinical assessments ABT-199 exhibited antitumor activity against hematologic malignancies including BCL-2-dependent non-Hodgkin lymphoma (NHL) and acute myeloid leukemia (AML) cell lines and xenograft models while demonstrating minimal platelet toxicity
Catalog No. 50-000-61953