Learn More
Apexbio Technology LLC ABT-263 (Navitoclax), 923564-51-6, MFCD12756219, 10 mg
SDP
Supplier:  Apexbio Technology LLC A300710
Encompass_Preferred
MF: C47H55ClF3N5O6S3, MW: 974.61, Synonyms: Navitoclax, ABT-263, ABT263, ABT 263. Solubility: 48.7305mg/mL in DMSO. ABT-263 is an orally sellecitive inhibitor of B-cell leukemia 2 (Bcl-2) family of proteins with potential antineoplastic activity.
Catalog No. 50-101-3827