Learn More
Medchemexpress LLC Abt-702 dihydrochloride | 1188890-28-9 | 99.6% | 536.25 | C22H21BrCl2N6O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1031611MG
Encompass_Preferred
ABT-702 dihydrochloride is a potent, non-nucleoside adenosine kinase (AK) inhibitor used in research to modulate adenosine signaling. It exhibits an IC50 of 1.7 nM against AK, has molecular formula C22H21BrCl2N6O, molecular weight 536.25, and CAS 1188890-28-9. Supplied in solid and DMSO solution formats for laboratory use.
  • Potent adenosine kinase inhibitor (IC50 1.7 nM)
  • High purity (99.64%)
  • Available as solid and 10 mM DMSO solution formats
  • Supplied for research use only
  • Suitable for biochemical and cell-based assays
Catalog No. 50-002-47690