Learn More
Medchemexpress LLC Abt-724 trihydrochloride | 587870-77-7 | 99.1% | 402.75 g/mol | C17H22Cl3N5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY103409100MG
Encompass_Preferred
ABT-724 trihydrochloride is a potent, selective dopamine D4 receptor agonist supplied for research use. It shows low-nanomolar activity at human D4 receptors and is used in pharmacological studies of dopaminergic signaling. The compound is provided as a solid, with defined purity and storage recommendations.
Catalog No. 50-002-46827