Learn More
Medchemexpress LLC HY-50907 10mg Medchemexpress, ABT-737 CAS:852808-04-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5090710MG
Encompass_Preferred
Medchemexpress, HY-50907 10mg ABT-737 CAS:852808-04-9 ABT-737 is a selective and BH3 mimetic Bcl-2, Bcl-xL and Bcl-w inhibitor with EC50s of 30.3 nM, 78.7 nM, and 197.8 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9353