Learn More
TARGETMOL CHEMICALS INC ABT-737 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T209925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABT-737 is a BH3 mimetic inhibitor of Bcl-xL Bcl-2 and Bcl-w (EC50s 78.7/30.3/197.8 nM). purity: 99%
Catalog No. 50-257-7227