Learn More
Selleck Chemical LLC ABT-751 (E7010) 10mM/1mL 141430-65-1
SDP
Supplier:  Selleck Chemical LLC S116510MM/1ML
Encompass_Preferred
ABT-751 (E7010) binds to the colchicine site on ?-tubulin and inhibits polymerization of microtubules, not a substrate for the MDR transporter and is active against cell lines resistant to vincristine, doxorubicin, and cisplatin. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0977