Learn More
Aat Bioquest ReadiUse™ ABTS Substrate Solution *Optimized for ELISA Assays with HRP Conjugates*
SDP
Supplier:  Aat Bioquest 11001
Encompass_Preferred
Horseradish peroxidase (HRP) and HRP conjugates facilitate the ABTS oxidation in the presence of hydrogen peroxide, turning ABTS into its blue-green oxidized product. This chromogenic reaction is widely used for quantify HRP in ELISA assays.
Catalog No. 50-195-3326