Learn More
Selleck Chemical LLC ABX464 S0076-25MG
SDP
Supplier:  Selleck Chemical LLC S007625MG
Encompass_Preferred
ABX464 (SPL-464) is a novel anti-HIV molecule that inhibits HIV-1 replication in stimulated peripheral blood mononuclear cells (PBMCs) from 5 different donors with IC50 ranging between 0 1 M and 0 5 M
Catalog No. 50-000-40293