Learn More
CPC Scientific H-Abz-Glu-Thr-Leu-Phe-Gln-Gly-Pro-Val-p-nitro-Phe-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HRV14001B
Encompass
ETLFQGPV corresponds to the 2C-3A cleavage site of the HRV type 14 protein. Abz-ETLFQGPVF(NO2)-NH2 is a FRET substrate for the HRV 3C protease in vitroSEQUENCE: H-Abz-Glu-Thr-Leu-Phe-Gln-Gly-Pro-Val-p-nitro-Phe-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Abz-ETLFQGPVF(NO2)-NH2MOLECULAR FORMULA: C57H77N13O16MOLECULAR WEIGHT: 1200.32STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [396096-53-0]SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:M.G.Cordingley et al., J. Biol. Chem., 265, 9062 (1990)SKU(s): HRV14-001A, HRV14-001B
Catalog No. 50-210-9576