Learn More
Medchemexpress LLC Abz-Nle-Lys-Arg-Arg-Ser-3-(NO2)Tyr acetate | 98.03% | 986.09 (free base) | 1 MG

Supplier: Medchemexpress LLC HYP5313A1MG
Abz-Nle-Lys-Arg-Arg-Ser-3-(NO2)Tyr acetate is a DEN protease substrate used in research related to dengue virus drug development. This product is a solid with a light yellow to yellow appearance, and has a purity of 98.03%.
- DEN protease substrate
- Used in research related to dengue virus drug development
- Molecular weight: 986.09 (free base)
- Formula: C43H67N15O12.xC2H4O2
- Sequence: Abz-Nle-Lys-Arg-Arg-Ser-{3-(NO2)Tyr}
- Storage as powder: -80°C for 2 years, -20°C for 1 year
- Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
- Solubility (in vitro): H2O: 10 mg/mL (ultrasonic needed)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.