Learn More
Medchemexpress LLC Abz-SDK(Dnp)P-OH | 860313-53-7 | 99.9% | 730.68 | 5 MG

Supplier: Medchemexpress LLC HYP101725MG
Abz-SDK(Dnp)P-OH is a fluorescence peptide that serves as a substrate for angiotensin I-converting enzyme (ACE). It features a fluorescent donor-acceptor pair, Abz and Dnp (2,4-dinitrophenyl).
- Fluorescence peptide
- Substrate of angiotensin I-converting enzyme (ACE)
- Contains fluorescent donor-acceptor pair Abz and Dnp (2,4-dinitrophenyl)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.