Learn More
Medchemexpress LLC Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp | 1042405-14-0 | 99.5% | 1285.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP10461100MG
Encompass_Preferred
Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp is a fluorescence-quenched peptide substrate designed for human proteinase 3 (PR3), exhibiting a kcat/Km of 1,570 mM-1s-1. It is utilized for detecting proteinase 3 (PR3) activity.
  • Fluorescence-quenched peptide substrate for human proteinase 3.
  • Can be used for the detection of proteinase 3 (PR3) activity.
  • High purity: 99.49%.
Catalog No. 50-004-24712