Learn More
Medchemexpress LLC Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp | 1042405-14-0 | 99.5% | 1285.33 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP10461
Encompass_Preferred
Abz-Val-Ala-Asp-Nva-Arg-Asp-Arg-Gln-EDDnp is a fluorescence-quenched peptide substrate for human proteinase 3 (kcat/Km = 1,570 mM-1s-1), utilized for detecting proteinase 3 (PR3) activity.
  • Fluorescence-quenched peptide substrate for human proteinase 3
  • Used for detection of proteinase 3 (PR3) activity
  • High purity of 99.49%
  • Stable for extended periods under recommended storage conditions
  • Available in powder or solvent formulations
Catalog No. 50-004-25456