Learn More
Medchemexpress LLC AC-261066 | 870773-76-5 | 99.27% | 353.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY108532100MG
Encompass_Preferred
AC-261066 is a potent, orally available, and isoform-selective retinoic acid beta2 (RARbeta2) receptor agonist with a pEC50 of 8.0. It exhibits good oral bioavailability in rats with an F oral of 52%.
  • Potent.
  • Orally available.
  • Isoform-selective retinoic acid beta2 (RARbeta2) receptor agonist.
  • Good oral bioavailability in rats.
Catalog No. 50-003-46081